Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dioctyl phthalate | 117-84-0 | 99.4% | 390.56 | 100 MG
SDP

Catalog No. 5000454712
Encompass_Preferred

Dioctyl phthalate (DNOP) is an active compound intended for research use only. It is not for sale to patients.

  • High purity: 99.4%
  • Appearance: Liquid, colorless to light yellow
  • Solubility: 100 mg/mL in DMSO
  • Storage: Store at room temperature, keep dry and cool. In solvent, store at -80°C for 1 year or -20°C for 6 months.

Catalog No. 50-004-54712 Supplier Medchemexpress LLC Supplier No. HYW013755100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dioctyl phthalate (DNOP) is an active compound intended for research use only. It is not for sale to patients.

  • High purity: 99.4%
  • Appearance: Liquid, colorless to light yellow
  • Solubility: 100 mg/mL in DMSO
  • Storage: Store at room temperature, keep dry and cool. In solvent, store at -80°C for 1 year or -20°C for 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.