Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dioctyl phthalate | 117-84-0 | 99.4% | 390.56 | 50 MG
SDP

Catalog No. 5000454448
Encompass_Preferred

Dioctyl phthalate (DNOP) is an active compound intended for research use only. This colorless to light yellow liquid has a high purity of 99.40% and a molecular weight of 390.56. It is crucial for various research applications.

  • High purity (99.40%)
  • Molecular weight of 390.56
  • Appears as a colorless to light yellow liquid
  • Suitable for research applications
  • Store at room temperature, keep dry and cool
  • Store in solvent at -80°C for 1 year or -20°C for 6 months

Catalog No. 50-004-54448 Supplier Medchemexpress LLC Supplier No. HYW01375550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dioctyl phthalate (DNOP) is an active compound intended for research use only. This colorless to light yellow liquid has a high purity of 99.40% and a molecular weight of 390.56. It is crucial for various research applications.

  • High purity (99.40%)
  • Molecular weight of 390.56
  • Appears as a colorless to light yellow liquid
  • Suitable for research applications
  • Store at room temperature, keep dry and cool
  • Store in solvent at -80°C for 1 year or -20°C for 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.