Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dioctyl phthalate (DNOP) | 117-84-0 | 99.4% | 390.56 | 250 MG
SDP

Catalog No. 5000366380
Encompass_Preferred

Dioctyl phthalate (DNOP) is an active compound presented as a colorless to light yellow liquid. It is recommended to store this product at room temperature, keeping it dry and cool. When dissolved in solvent, it can be stored at -80°C for one year or -20°C for six months.

  • Synonyms: DNOP, di-n-octyl phthalate, phthalic acid dioctyl ester
  • Purity: 99.40%
  • Molecular weight: 390.56
  • Molecular formula: C24H38O4
  • CAS number: 117-84-0
  • Appearance: Liquid, colorless to light yellow
  • In vitro solubility: DMSO: 100 mg/mL (needs ultrasonic)
  • In vivo solubility: 10% DMSO, 90% corn oil; ≥ 2.5 mg/mL (clear solution)

Catalog No. 50-003-66380 Supplier Medchemexpress LLC Supplier No. 250MGHYW013755
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dioctyl phthalate (DNOP) is an active compound presented as a colorless to light yellow liquid. It is recommended to store this product at room temperature, keeping it dry and cool. When dissolved in solvent, it can be stored at -80°C for one year or -20°C for six months.

  • Synonyms: DNOP, di-n-octyl phthalate, phthalic acid dioctyl ester
  • Purity: 99.40%
  • Molecular weight: 390.56
  • Molecular formula: C24H38O4
  • CAS number: 117-84-0
  • Appearance: Liquid, colorless to light yellow
  • In vitro solubility: DMSO: 100 mg/mL (needs ultrasonic)
  • In vivo solubility: 10% DMSO, 90% corn oil; ≥ 2.5 mg/mL (clear solution)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.