Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dioctyl sulfosuccinate sodium salt

Catalog No. 3235861KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-358-61KG 1 kg
32-358-6100G 100 g
32-358-625G 25 g
32-358-6500G 500 g
32-358-65KG 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 32-358-61KG Supplier Sigma Aldrich Supplier No. 3235861KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dioctyl sulfosuccinate sodium salt

Specifications

CAS 577-11-7
Melting Point 173°C to 179°C (lit.)
Molecular Formula C20H37NaO7S
Linear Formula CH3(CH2)3 CH(C2H5)CH2O2CCH2CH(SO3Na)CO2CH2CH(C2H5)(CH2)3 CH3
MDL Number MFCD00012455
Quantity 1 kg
Synonym AOT; Bis(2-ethylhexyl) sulfosuccinate sodium salt; DOSS; Docusate sodium
Molecular Weight (g/mol) 444.56
Percent Purity ≥97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.