Learn More
National Diagnostics Diogenes Cellular Luminescence Enhancement System, Superoxide detection, Non-cytotoxic, Requires fewer cells, Assay Yield: Up to 100 assays per 10 ml
SDP
Catalog No. NC3724502
Encompass
Diogenes Cellular Luminescence Enhancement System, Superoxide detection, Non-cytotoxic, Requires fewer cells, Assay Yield: Up to 100 assays per 10 ml
Catalog No. NC3724502 Supplier National Diagnostics Supplier No. CL202C
Add to Cart
Add to Cart
Request a Quote
Diogenes Cellular Luminescence Enhancement System, Superoxide detection, Non-cytotoxic, Requires fewer cells, Assay Yield: Up to 100 assays per 10 ml