Learn More
YMC America, Inc Diol col 200A 5um 50x1.0mm
SDP
Catalog No. 5000879819
Encompass_Preferred
YMC-Pack Diol is a size exclusion chromatography column packed with dihydroxypropyl-bonded silica and available in four different pore sizes It exhibits high mechanical strength and is useful for molecular weight determination of proteins and sugars The 2 m and 3 m particle sizes provide high resolution and fast analysis
Catalog No. 50-008-79819 Supplier YMC America, Inc Supplier No. DL20S050501WT
Add to Cart
Add to Cart
YMC-Pack Diol is a size exclusion chromatography column packed with dihydroxypropyl-bonded silica and available in four different pore sizes It exhibits high mechanical strength and is useful for molecular weight determination of proteins and sugars The 2 m and 3 m particle sizes provide high resolution and fast analysis