Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc DIOL-GFC 5UM 300A 500X20MM
SDP

Catalog No. 5090005414
Encompass_Preferred

Diol-Gfc 5Um; 300A 500 X 20mm

Catalog No. 50-900-05414 Supplier YMC America, Inc Supplier No. DL30S055020WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Diol-Gfc 5Um; 300A 500 X 20mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.