Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc DIOL-GFC 5UM 60A 50X20MM
SDP

Supplier:  YMC America, Inc DL06S050520WTG

Encompass_Preferred

Diol-Gfc 5Um; 60A 50 X 20mm

Catalog No. 50-900-05381


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.