Learn More
YMC America, Inc Diol guard 60A 5um 20x2.1mm
SDP
Catalog No. 5000879917
Encompass
YMC-Pack Diol is a size exclusion chromatography column packed with dihydroxypropyl-bonded silica and available in four different pore sizes It exhibits high mechanical strength and is useful for molecular weight determination of proteins and sugars The 2 m and 3 m particle sizes provide high resolution and fast analysis
Catalog No. 50-008-79917 Supplier YMC America, Inc Supplier No. DL06S0502Q1GC
Add to Cart
Add to Cart
YMC-Pack Diol is a size exclusion chromatography column packed with dihydroxypropyl-bonded silica and available in four different pore sizes It exhibits high mechanical strength and is useful for molecular weight determination of proteins and sugars The 2 m and 3 m particle sizes provide high resolution and fast analysis