Learn More
YMC America, Inc Diol-HG col 120A 10um 250x4mm
SDP
Catalog No. 5000880940
Encompass_Preferred
YMC-Pack Diol is a size exclusion chromatography column packed with dihydroxypropyl-bonded silica and available in four different pore sizes It exhibits high mechanical strength and is useful for molecular weight determination of proteins and sugars The 2 m and 3 m particle sizes provide high resolution and fast analysis
Catalog No. 50-008-80940 Supplier YMC America, Inc Supplier No. DLG12S112504WT
YMC-Pack Diol is a size exclusion chromatography column packed with dihydroxypropyl-bonded silica and available in four different pore sizes It exhibits high mechanical strength and is useful for molecular weight determination of proteins and sugars The 2 m and 3 m particle sizes provide high resolution and fast analysis