Learn More
YMC America, Inc Diol-NP 120A 5um 150x.5mm 1/16
SDP
Catalog No. 5000879323
Encompass
YMC-Pack Diol-NP is a versatile alternative to silica for normal phase separations and provides much better reproducibility when compared to bare silica material It shows typical normal phase retention behavior when used with low-polarity mobile phases due to hydroxyl groups on the surface of the stationary phase acting as polar groups YMC-Pack Diol-NP can also be used in HILIC-mode with organic/water mobile phases
Catalog No. 50-008-79323 Supplier YMC America, Inc Supplier No. DN12S0515J0AU
Add to Cart
Add to Cart
YMC-Pack Diol-NP is a versatile alternative to silica for normal phase separations and provides much better reproducibility when compared to bare silica material It shows typical normal phase retention behavior when used with low-polarity mobile phases due to hydroxyl groups on the surface of the stationary phase acting as polar groups YMC-Pack Diol-NP can also be used in HILIC-mode with organic/water mobile phases