Learn More
YMC America, Inc Diol-NP 60A 5um 75x0.5mm 1/16
SDP
Catalog No. 5000878669
Encompass
YMC-Pack Diol-NP is a versatile alternative to silica for normal phase separations and provides much better reproducibility when compared to bare silica material It shows typical normal phase retention behavior when used with low-polarity mobile phases due to hydroxyl groups on the surface of the stationary phase acting as polar groups YMC-Pack Diol-NP can also be used in HILIC-mode with organic/water mobile phases
Catalog No. 50-008-78669 Supplier YMC America, Inc Supplier No. DN06S05L5J0AU
YMC-Pack Diol-NP is a versatile alternative to silica for normal phase separations and provides much better reproducibility when compared to bare silica material It shows typical normal phase retention behavior when used with low-polarity mobile phases due to hydroxyl groups on the surface of the stationary phase acting as polar groups YMC-Pack Diol-NP can also be used in HILIC-mode with organic/water mobile phases