Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diolein, > 99%, MP Biomedicals™

Catalog No. ICN19515180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19515180 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19515180 Supplier MP Biomedicals, Inc Supplier No. 0219515180
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25637-84-7
Molecular Formula C39H72O5
Synonym Glycerol-1, 2-and-1, 3-dioleate

Specifications

Color Colorless
Density 0.921g/mL
Assay Percent Range ∽ 99%
Quantity 100 mg
Formula Weight 621
Percent Purity >99%
Physical Form Frozen Liquid
Chemical Name or Material Diolein
EINECSNumber 247-144-2
Recommended Storage Store at - 20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.