Learn More
Abcam 1,2-Dioleoyl-3-(pyren-1-yl) decanoyl-rac-glycerol, 1MG
Catalog No. 50996093
Encompass_Preferred
MW 975.5 Da. Fluorescent triglyceride analog useful for measuring cholesteryl ester transfer and protein-mediated triacylglycerol transport between plasma lipoproteins. Popular substrate for detections of lipoprotein lipase and hepatic triacylglycerol lipases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-093 Supplier Abcam Supplier No. AB2754921MG
MW 975.5 Da. Fluorescent triglyceride analog useful for measuring cholesteryl ester transfer and protein-mediated triacylglycerol transport between plasma lipoproteins. Popular substrate for detections of lipoprotein lipase and hepatic triacylglycerol lipases.
The product is subject to the following: Abcam Restricted Use Statement