Learn More
Medchemexpress LLC Dioleoylphosphatidylethanolamine | 4004-05-1 | 100.0% | 744.03 | 250 MG
SDP
Catalog No. 5000439516
Encompass_Preferred
Dioleoylphosphatidylethanolamine (DOPE) is a neutral helper lipid primarily utilized in the formation of cationic liposomes. It is designed to combine with cationic phospholipids to significantly enhance the transfection efficiency of naked siRNA.
  • Used as a neutral helper lipid for cationic liposomes.
  • Enhances transfection efficiency of naked siRNA when combined with cationic phospholipids.
  • Participates in liposome-containing formulations exhibiting fusogenicity.
  • Supports siRNA knock-down effects on macrophages and dendritic cells when encapsulated in LNPs.
  • Functions as a potential efficient siRNA delivery system for murine and human macrophages and dendritic cells.
Catalog No. 50-004-39516 Supplier Medchemexpress LLC Supplier No. 250MGHY112005
Dioleoylphosphatidylethanolamine (DOPE) is a neutral helper lipid primarily utilized in the formation of cationic liposomes. It is designed to combine with cationic phospholipids to significantly enhance the transfection efficiency of naked siRNA.
  • Used as a neutral helper lipid for cationic liposomes.
  • Enhances transfection efficiency of naked siRNA when combined with cationic phospholipids.
  • Participates in liposome-containing formulations exhibiting fusogenicity.
  • Supports siRNA knock-down effects on macrophages and dendritic cells when encapsulated in LNPs.
  • Functions as a potential efficient siRNA delivery system for murine and human macrophages and dendritic cells.