Learn More
Cayman Chemical Dioleyldimethylammonium (chloride), Purity: ≥95%, CAS: 7212-69-3, 10 mg
SDP
Catalog No. 502917064
Encompass_Preferred
A cationic lipid; has been used as a transfection reagent; liposomes containing DODAC and DOPE and encapsulating a plasmid DNA reporter induce reporter gene expression in C2C12 mouse skeletal muscle myoblasts
Catalog No. 50-291-7064 Supplier Cayman Chemical Supplier No. 3879710 MG
A cationic lipid; has been used as a transfection reagent; liposomes containing DODAC and DOPE and encapsulating a plasmid DNA reporter induce reporter gene expression in C2C12 mouse skeletal muscle myoblasts