Learn More
YMC America, Inc DiolNPHG 120A 50um 100x10mm
SDP
Catalog No. 5000880349
Encompass_Preferred
YMC-Pack Diol-NP is a versatile alternative to silica for normal phase separations and provides much better reproducibility when compared to bare silica material It shows typical normal phase retention behavior when used with low-polarity mobile phases due to hydroxyl groups on the surface of the stationary phase acting as polar groups YMC-Pack Diol-NP can also be used in HILIC-mode with organic/water mobile phases
Catalog No. 50-008-80349 Supplier YMC America, Inc Supplier No. DNG12S501010WT
YMC-Pack Diol-NP is a versatile alternative to silica for normal phase separations and provides much better reproducibility when compared to bare silica material It shows typical normal phase retention behavior when used with low-polarity mobile phases due to hydroxyl groups on the surface of the stationary phase acting as polar groups YMC-Pack Diol-NP can also be used in HILIC-mode with organic/water mobile phases