Learn More
Thermo Scientific Deuterium Lamp for Thermo Scientific Dionex HPLC Systems [Quantity: 1]
Catalog No. NC0373589 Shop All Thermo Scientific Products
Encompass
A replacement deuterium lamp emitting a 190-380nm UV spectrum for UV detection in liquid chromatography systems.
Catalog No. NC0373589 Supplier Thermo Scientific Supplier No. 066347
A replacement deuterium lamp emitting a 190-380nm UV spectrum for UV detection in liquid chromatography systems.