Learn More
Thermo Scientific Dionex™ AS29 Eluent Concentrate, Sodium Carbonate/Bicarbonate Concentrate (100x)
Catalog No. NC1498675 Shop All Thermo Scientific Products
Encompass
Save time and reduce preparation errors and contamination that may occur during eluent preparation with the ready-to-dilute Thermo Scientific™ Dionex™ AS29 Eluent Concentrate. For use with Dionex IonPac AS29-Fast-4µm IC columns, simply prepare eluent by diluting the concentrate in deionized water. The 250mL bottle of 100x concentrate is made with only high purity, reagent grade chemicals and water (18.2 MΩ-cm), and quality checked for ionic contamination. The concentrate contains verified concentrations of 0.45M carbonate and 0.20M bicarbonate.
Catalog No. NC1498675 Supplier Thermo Scientific Supplier No. 302952
Save time and reduce preparation errors and contamination that may occur during eluent preparation with the ready-to-dilute Thermo Scientific™ Dionex™ AS29 Eluent Concentrate. For use with Dionex IonPac AS29-Fast-4µm IC columns, simply prepare eluent by diluting the concentrate in deionized water. The 250mL bottle of 100x concentrate is made with only high purity, reagent grade chemicals and water (18.2 MΩ-cm), and quality checked for ionic contamination. The concentrate contains verified concentrations of 0.45M carbonate and 0.20M bicarbonate.