Learn More
Thermo Scientific Dionex Asrs300 Suppressor 2Mm
Catalog No. NC1268660 Shop All Thermo Scientific Products
Encompass
Electrolytically regenerated eluent conductivity suppressor for ion chromatography systems compatible with 2MM columns and 0.5ML/min flow constructed of PEEK™ polymer.
Catalog No. NC1268660 Supplier Thermo Scientific Supplier No. SP6948
Electrolytically regenerated eluent conductivity suppressor for ion chromatography systems compatible with 2MM columns and 0.5ML/min flow constructed of PEEK™ polymer.