Learn More
Thermo Scientific™ Dionex™ EG Eluent Generator Kits
Catalog No. p-6145597
Encompass
Change view
Click to view available options
Description:
Analytical EG Kit
For Use With (Equipment):
Dionex ICS-6000 EG Eluent Generator
Catalog No. Description For Use With (Equipment)
50-134-5955 Analytical EG Kit Dionex ICS-6000 EG Eluent Generator
1 options
Catalog No. 50-134-5955 Supplier Thermo Scientific™ Supplier No. 075522
Only null left
Add to Cart
Add to Cart
Run ion chromatography gradients as easily as isocratic by installing the Thermo Scientific™ Dionex™ EG Cartridge Kits in the Dionex ICS-6000 EG Eluent Generator.
Achieve reproducible and reliable chromatography separations using Thermo Scientific™ Dionex™ Reagent-Free IC (RFIC™) technologies. Dionex Eluent Generator accessories allow you to operate your RFIC system with minimal effort through plug-and-play setup.
Operate your ion chromatography system with minimal effort when using ThermoScientific™ Dionex™ EG Eluent Generator accessories.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Dionex ICS-6000 EG Eluent Generator
Type Analytical Eluent Generator Kit (Eluent Concentration: 0.01 to 100 mM)
Flow Rate 0.1 to 3.000 mL/min.
Pressure 5000 psi
Solvents Max. Solvent concentration - Cations: None; Anions: 25%
Amperage 90-265 VAC 47-63 Hz
Includes Analytical degasser module and tubing
Description Analytical EG Kit