Learn More
Analytical Sales & Services Dionex ICS 900 1000 1100 15
Catalog No. 5000804233
Encompass
Dionex ICS 900 1000 1100 1500 1600 2000 3000 Outlet Check Valve Assembly PEEK; OEM No. 45721
Qty/Pkg: EACH
Catalog No. 50-008-04233 Supplier Analytical Sales & Services Supplier No. 925047
Dionex ICS 900 1000 1100 1500 1600 2000 3000 Outlet Check Valve Assembly PEEK; OEM No. 45721
Qty/Pkg: EACH