Learn More
Thermo Scientific Dionex IonPac AS25 IC Columns | | 1 EA
Supplier:  Thermo Scientific 076014
Encompass
- High capacity 350eq per column 4x250mm- Ideal for polyvalent anions including polysulfates polyphosphates and polycarboxylates- Ultralow hydrophobicity allows fast analysis of polarizable anions- Low background signal and enhanced analyte sensitivity with the Thermo Scientific Dionex Anion Self-Regenerating Suppressor ASRS 300 and a Reagent-Free RFIC system- Compatible with organic solvents to enhance analyte solubility modify column selectivity and effectively clean the column
Catalog No. NC0458152