Learn More
Thermo Scientific Dionex IonPac CS16-4 µm column
Catalog No. NC1752068 Shop All Thermo Scientific Products
Encompass
Type Standard Bore Guard | Flow Rate 0.64 to 1.0 mL/min. | Packing Material Ethylvinylbenzene/Divinylbenzene | Length (Metric) 50 mm | Particle Size 4 µm | Diameter (Metric) 4 mm | Max. Pressure 5000 psi (345 bar) | For Use With (Application) Cation Exchange | Column Capacity 1070 µeq | Crosslinking 55% DVB | Column Construction PEEK | Column Chemistry Acidic | Hydrophobicity Medium | Mobile Phase Compatibility Acidic Eluents (pH 0 to 7), 0 to 100% Acetonitrile, 0 to 100% Acetone, Alcohols Should be Avoided | Ion Exchange Group Carboxylic Acid | For Use With (Equipment) Dionex IonPac CS16-4 µm Standard Bore Separator Column | Stationary Phase IonPac CS16-4 µm
Catalog No. NC1752068 Supplier Thermo Scientific Supplier No. 088585
Type Standard Bore Guard | Flow Rate 0.64 to 1.0 mL/min. | Packing Material Ethylvinylbenzene/Divinylbenzene | Length (Metric) 50 mm | Particle Size 4 µm | Diameter (Metric) 4 mm | Max. Pressure 5000 psi (345 bar) | For Use With (Application) Cation Exchange | Column Capacity 1070 µeq | Crosslinking 55% DVB | Column Construction PEEK | Column Chemistry Acidic | Hydrophobicity Medium | Mobile Phase Compatibility Acidic Eluents (pH 0 to 7), 0 to 100% Acetonitrile, 0 to 100% Acetone, Alcohols Should be Avoided | Ion Exchange Group Carboxylic Acid | For Use With (Equipment) Dionex IonPac CS16-4 µm Standard Bore Separator Column | Stationary Phase IonPac CS16-4 µm