Description
Thermo Scientific™ Ultra-Low Dead Volume Union is for nano electrospray ionization.
- Dead volume of <7nL
- With two nuts and two ferrules
- I.D.: 1/16in.
Specifications
Specifications
| Description | Union |
| For Use With (Equipment) | Nano ES |
| Diameter (English) Outer | 1/16 in. |
| Product Line | nanoLC |