Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-DIOP, 98%, Thermo Scientific™

Catalog No. AAB21029MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21029MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB21029MD Supplier Thermo Scientific Chemicals Supplier No. B21029MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32305-98-9
Molecular Formula C31H32O2P2
Molecular Weight (g/mol) 498.54
MDL Number MFCD00014107
InChI Key VCHDBLPQYJAQSQ-UHFFFAOYNA-N
Synonym --diop, r,r-diop, unii-esk57w53ci, esk57w53ci, 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl bis methylene bis diphenylphosphine, --2,3-o-isopropylidene-2,3-dihydroxy-1,4-bis diphenylphosphino butane, 4r,5r---4,5-bis diphenylphosphinomethyl-2,2-dimethyl-1,3-dioxolane, 2r,3r---1,4-bis diphenylphosphino-2,3-o-isopropylidene-2,3-butanediol, 4r,5r---bis diphenylphosphinomethyl-2,2-dimethyl-1,3-dioxolane, --1,4-bis diphenylphosphino-1,4-dideoxy-2,3-o-isopropylidene-l-threitol
PubChem CID 395120
IUPAC Name [(4R,5R)-5-(diphenylphosphanylmethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl-diphenylphosphane
SMILES CC1(C)OC(CP(C2=CC=CC=C2)C2=CC=CC=C2)C(CP(C2=CC=CC=C2)C2=CC=CC=C2)O1

Specifications

Melting Point 88°C to 90°C
Quantity 250 mg
Beilstein 1441045
Sensitivity Air sensitive
Optical Rotation −12.5° (c=5 in Benzene)
Formula Weight 498.55
Percent Purity 98%
Chemical Name or Material (-)-DIOP
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 250-984-2
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.