Learn More
Cayman Chemical Dios-Arg (trifluoroacetate salt), Purity: ≥95%, CAS: 1807353-31-6, 10 mg
SDP
Catalog No. 502936270
Encompass_Preferred
A steroid-based cationic lipid; forms a complex with plasmid DNA and decreases plasmid DNA migration in an agarose-gel retardant assay at charge ratio ≥4; facilitates transfection of plasmid DNA into H1299 and HeLa cells; has been used, coupled to DOPE, to bind siRNA and plasmid DNA to form cationic lipid nanoparticles for intracellular transport
Catalog No. 50-293-6270 Supplier Cayman Chemical Supplier No. 2594510 MG
A steroid-based cationic lipid; forms a complex with plasmid DNA and decreases plasmid DNA migration in an agarose-gel retardant assay at charge ratio ≥4; facilitates transfection of plasmid DNA into H1299 and HeLa cells; has been used, coupled to DOPE, to bind siRNA and plasmid DNA to form cationic lipid nanoparticles for intracellular transport