Learn More
Apexbio Technology LLC Dioscin, 20mg. Cas: 19057-60-4 MFCD: MFCD02094174. Natural saponin with anti-tumor activities
SDP
Supplier:  Apexbio Technology LLC N172320
Encompass_Preferred
Extracted from (Dioscorea nipponica Makino); Suitability:Methanol,ethanol,acetic acid,slightly soluble in acetone,amyl alcohol;Store the product in sealed,cool and dry condition Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5642