Learn More
Medchemexpress LLC Diosgenin glucoside | 14144-06-0 | MFCD00867497 | 576.76 | 1 MG
SDP
Catalog No. 5000415364
Encompass_Preferred
Diosgenin glucoside is a saponin compound extracted from Trillium tschonoskii. It provides neuroprotection by regulating microglial M1 polarization and protects against spinal cord injury by regulating autophagy and alleviating apoptosis. It is for research use only.
  • Supplied as a white to off-white solid
  • Provides neuroprotection by regulating microglial M1 polarization
  • Protects against spinal cord injury
  • Promotes motor function recovery and reduces tissue damage after spinal cord injury in rat models
  • Reduces cell apoptosis and activates autophagy in SCI rat models
Catalog No. 50-004-15364 Supplier Medchemexpress LLC Supplier No. HYN07301MG
Diosgenin glucoside is a saponin compound extracted from Trillium tschonoskii. It provides neuroprotection by regulating microglial M1 polarization and protects against spinal cord injury by regulating autophagy and alleviating apoptosis. It is for research use only.
  • Supplied as a white to off-white solid
  • Provides neuroprotection by regulating microglial M1 polarization
  • Protects against spinal cord injury
  • Promotes motor function recovery and reduces tissue damage after spinal cord injury in rat models
  • Reduces cell apoptosis and activates autophagy in SCI rat models