Learn More
TARGETMOL CHEMICALS INC DIOSGENIN GLUCOSIDE 25MG
SDP
Catalog No. 502580087
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Diosgenin glucoside (Trillin) and other synthetic glycosides with similar activities may be of use in the management of hypercholesterolemia and atherosclerosis. Significant inhibition of Diosgenin glucoside synthesis and stimulation of solasodine glucosylation was found only with PC molecular species containing fatty acids with a chain length of 12-18 carbon atoms. purity: 99%
Catalog No. 50-258-0087 Supplier TARGETMOL CHEMICALS INC Supplier No. T387425MG
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Diosgenin glucoside (Trillin) and other synthetic glycosides with similar activities may be of use in the management of hypercholesterolemia and atherosclerosis. Significant inhibition of Diosgenin glucoside synthesis and stimulation of solasodine glucosylation was found only with PC molecular species containing fatty acids with a chain length of 12-18 carbon atoms. purity: 99%