Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diosgenin

Catalog No. AAJ6097614
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6097614 25 g
AAJ6097606 5 g
AAJ6097622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6097614 Supplier Thermo Scientific Chemicals Supplier No. J6097614
  • $166.80
    $141.65 / Each of 1

    Save $25.15

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 512-04-9 | C27H42O3 | 414.63 g/mol

Diosgenin is the precursor for the semisynthesis of progesterone which in turn was used in early combined oral contraceptive pills. The unmodified steroid has estrogenic activity and can reduce the level of serum cholesterol. Diosgenin has been used in traditional medicine as an antihypercholesterolemia, antihypertriacylglycerolemia, antidiabetes and antihyperglycemia agent. Diosgenin is a steroid sapogenin used in the semi-synthesis of pregnenolone, cortisone, and progesterone. Diosgenin has estrogenic activity and can reduce the level of serum cholesterol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diosgenin is the precursor for the semisynthesis of progesterone which in turn was used in early combined oral contraceptive pills. The unmodified steroid has estrogenic activity and can reduce the level of serum cholesterol. Diosgenin has been used in traditional medicine as an antihypercholesterolemia, antihypertriacylglycerolemia, antidiabetes and antihyperglycemia agent. Diosgenin is a steroid sapogenin used in the semi-synthesis of pregnenolone, cortisone, and progesterone. Diosgenin has estrogenic activity and can reduce the level of serum cholesterol.

Solubility
Soluble in chloroform (50 mg/ml), ethanol (83 mg/ml at 25°C), water (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512-04-9
Molecular Formula C27H42O3
Molecular Weight (g/mol) 414.63
MDL Number MFCD00016887,MFCD01002142
InChI Key WQLVFSAGQJTQCK-VKROHFNGSA-N
Synonym 3-beta-Hydroxy-5-spirostene; Nitogenin
PubChem CID 46783391
IUPAC Name (1'S,2R,2'S,4'S,5R,7'S,8'R,9'S,12'S,13'R,16'S)-5,7',9',13'-tetramethyl-5'-oxaspiro[oxane-2,6'-pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan]-18'-en-16'-ol
SMILES C[C@H]1[C@H]2[C@H](C[C@H]3[C@@H]4CC=C5C[C@@H](O)CC[C@]5(C)[C@H]4CC[C@]23C)O[C@]11CC[C@@H](C)CO1

Specifications

Melting Point 200°C to 206°C
Color White
Quantity 25 g
Beilstein 94582
Merck Index 14,3295
Solubility Information Soluble in chloroform (50mg/ml),ethanol (83mg/ml at 25°C),water (<1mg/ml at 25°C),DMSO (<1mg/ml at 25°C),and methanol.
Formula Weight 414.63
Physical Form Powder
Chemical Name or Material Diosgenin
EINECSNumber 208-134-3
RTECSNumber WH1450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.