Learn More
Medchemexpress LLC HY-N0125 50mg Medchemexpress, Diosmetin CAS:520-34-3 Purity:>98%
SDP
Catalog No. 501933411
Encompass_Preferred
Medchemexpress, HY-N0125 50mg Diosmetin CAS:520-34-3 Diosmetin is a natural flavonoid which inhibits human CYP1A enzyme activity with an IC 50 of 40 μM in HepG2 cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3411 Supplier Medchemexpress LLC Supplier No. HYN012550MG
Medchemexpress, HY-N0125 50mg Diosmetin CAS:520-34-3 Diosmetin is a natural flavonoid which inhibits human CYP1A enzyme activity with an IC 50 of 40 μM in HepG2 cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.