Learn More
TARGETMOL CHEMICALS INC DiosMetin 7-O-beta-D-Glucuroni
SDP
Catalog No. 502879495
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. DiosMetin 7-O-beta-D-Glucuronide, an antioxidant constituent found in the fruits of Luffa cylindrical, supplies antioxidant benefits when sponge gourds are consumed. Purity 99.47%
Catalog No. 50-287-9495 Supplier TARGETMOL CHEMICALS INC Supplier No. T110475MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. DiosMetin 7-O-beta-D-Glucuronide, an antioxidant constituent found in the fruits of Luffa cylindrical, supplies antioxidant benefits when sponge gourds are consumed. Purity 99.47%