Learn More
Cayman Chemical Diosmetin-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1189728-54-8, 1 mg
SDP
Catalog No. 502988865
Encompass_Preferred
An internal standard for the quantification of diosmetin by GC- or LC-MS
Catalog No. 50-298-8865 Supplier Cayman Chemical Supplier No. 301221 MG
An internal standard for the quantification of diosmetin by GC- or LC-MS