Learn More
Cayman Chemical DIosmetIn-d3 500ug
SDP
Catalog No. 502989865
Encompass_Preferred
An internal standard for the quantification of diosmetin by GC- or LC-MS
Catalog No. 50-298-9865 Supplier Cayman Chemical Supplier No. 30122500 UG
An internal standard for the quantification of diosmetin by GC- or LC-MS