Learn More
Selleck Chemical LLC Diosmin S2292-200mg
SDP
Catalog No. 503112688
Encompass_Preferred
Diosmin is an agonist of the aryl hydrocarbon receptor (AhR) Diosmin is a semisynthetic phlebotropic agent and a flavonoid found in a variety of citrus fruits
Catalog No. 50-311-2688 Supplier Selleck Chemical LLC Supplier No. S2292200MG
Diosmin is an agonist of the aryl hydrocarbon receptor (AhR) Diosmin is a semisynthetic phlebotropic agent and a flavonoid found in a variety of citrus fruits