Learn More
Medchemexpress LLC HY-N0178 50mg Medchemexpress, Diosmin CAS:520-27-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN017850MG
Encompass_Preferred
Medchemexpress, HY-N0178 50mg Diosmin CAS:520-27-4 Diosmin is a flavonoid found in a variety of citrus fruits and also an agonist of the aryl hydrocarbon receptor ( AhR ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3449