Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diosmin | 520-27-4 | 97.5% | 608.54 g/mol | C28H32O15 | 10 MG
SDP

Catalog No. 5000295879
Encompass_Preferred

Diosmin is an analytical reference standard of the flavonoid diosmin supplied for research and analytical applications. It is intended for assay development, method validation, and chromatographic analysis.

  • Purity 97.54% (analytical grade).
  • Cas number 520-27-4 for unambiguous identification.
  • Molecular formula C28H32O15 and molecular weight 608.54 g/mol.
  • Suitable for HPLC, LC-MS, and other analytical workflows.
  • Available in small pack sizes, such as 10 MG, for reference use.

Catalog No. 50-002-95879 Supplier Medchemexpress LLC Supplier No. HYN0178R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diosmin is an analytical reference standard of the flavonoid diosmin supplied for research and analytical applications. It is intended for assay development, method validation, and chromatographic analysis.

  • Purity 97.54% (analytical grade).
  • Cas number 520-27-4 for unambiguous identification.
  • Molecular formula C28H32O15 and molecular weight 608.54 g/mol.
  • Suitable for HPLC, LC-MS, and other analytical workflows.
  • Available in small pack sizes, such as 10 MG, for reference use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.