Learn More
Dioxins and Furans in Tissue - PT, MilliporeSigma™ Supelco™
Catalog No. 111017293 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
Catalog No. Quantity
11-101-7293 10 g
1 options
Catalog No. 11-101-7293 Supplier MilliporeSigma Supelco Supplier No. SPE016TIS10G
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Featured Industries: Environmental
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
CAS 1746-01-6
Type Proficiency Testing Material
Quantity 10 g