Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dioxopromethazine hydrochloride | 15374-15-9 | MFCD01706903 | 100.0% | 352.88 | C17H21ClN2O2S | 25 MG
SDP

Catalog No. 5000242757
Encompass_Preferred

Dioxopromethazine hydrochloride is the hydrochloride salt of a phenothiazine-derived antihistamine used in research. It is orally active and reported to inhibit asthmatic symptoms; the material is supplied for research use only and is presented as a white to off-white solid in small, bench-scale quantities.

  • Orally active antihistamine suitable for respiratory and histamine receptor research.
  • High purity (reported as 99.97%) for consistent experimental results.
  • Molecular formula C17H21ClN2O2S and molecular weight 352.88.
  • White to off-white solid appearance, compatible with common laboratory handling.
  • Available in small quantity formats for bench-scale work.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months).

Catalog No. 50-002-42757 Supplier Medchemexpress LLC Supplier No. HY107787A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dioxopromethazine hydrochloride is the hydrochloride salt of a phenothiazine-derived antihistamine used in research. It is orally active and reported to inhibit asthmatic symptoms; the material is supplied for research use only and is presented as a white to off-white solid in small, bench-scale quantities.

  • Orally active antihistamine suitable for respiratory and histamine receptor research.
  • High purity (reported as 99.97%) for consistent experimental results.
  • Molecular formula C17H21ClN2O2S and molecular weight 352.88.
  • White to off-white solid appearance, compatible with common laboratory handling.
  • Available in small quantity formats for bench-scale work.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.