Learn More
Medchemexpress LLC Dioxopromethazine hydrochloride | 15374-15-9 | MFCD01706903 | 100.0% | 352.88 g·mol⁻¹ | C17H21ClN2O2S | 5 MG
SDP
Catalog No. 5000240271
Encompass_Preferred
Dioxopromethazine hydrochloride is an orally active antihistamine used in research to study histamine-related responses and the inhibition of asthmatic symptoms. Supplied as the hydrochloride salt, it is intended for preclinical and in vitro assay applications.
Catalog No. 50-002-40271 Supplier Medchemexpress LLC Supplier No. HY107787A5MG
Dioxopromethazine hydrochloride is an orally active antihistamine used in research to study histamine-related responses and the inhibition of asthmatic symptoms. Supplied as the hydrochloride salt, it is intended for preclinical and in vitro assay applications.