Learn More
Medchemexpress LLC Dioxopromethazine hydrochloride | 15374-15-9 | MFCD01706903 | 100.0% | 352.88 g·mol⁻¹ | C17H21ClN2O2S | 50 MG
SDP
Catalog No. 5000241215
Encompass_Preferred
Dioxopromethazine hydrochloride is a phenothiazine-derived, orally active antihistamine used in preclinical research to study histamine-mediated responses and to investigate inhibition of asthmatic symptoms. Supplied as a hydrochloride salt with high reported purity, it is intended for laboratory research use only.
  • Orally active compound suitable for in vivo and in vitro research.
  • Modulates histamine-related pathways and may inhibit asthmatic symptoms.
  • Provided as a hydrochloride salt to aid solubility in polar solvents.
  • High reported purity appropriate for research applications.
  • Available with safety data sheet and technical documentation for laboratory handling.
Catalog No. 50-002-41215 Supplier Medchemexpress LLC Supplier No. HY107787A50MG
Dioxopromethazine hydrochloride is a phenothiazine-derived, orally active antihistamine used in preclinical research to study histamine-mediated responses and to investigate inhibition of asthmatic symptoms. Supplied as a hydrochloride salt with high reported purity, it is intended for laboratory research use only.
  • Orally active compound suitable for in vivo and in vitro research.
  • Modulates histamine-related pathways and may inhibit asthmatic symptoms.
  • Provided as a hydrochloride salt to aid solubility in polar solvents.
  • High reported purity appropriate for research applications.
  • Available with safety data sheet and technical documentation for laboratory handling.