Learn More
Binder Inc O₂ control range
SDP
Supplier:  Binder Inc 86120028
Encompass_Preferred
O₂ control range - alternative to the standard range, for controlling O₂ concentrations between 10 and 90 vol. %, only in conjunction with option models with O₂ control
Catalog No. 86-120-028