Learn More
Med Vet International Dip Quick StainFixative, 500mL, J0322A1
Catalog No. 5000581501
Encompass_Preferred
JorVet Dip Quick Stain is excellent for use in blood cell differential counts or as a general cytology stain, and gives coloration similar to the standard Wright-Giemsa stain. The rapid 3-step procedure can be done right in the wide-mouth 180ml. container in 15 seconds. Fixative Only. 500mlNOTE: These components may interchange with other brands of 3-component differenential stains
Catalog No. 50-005-81501 Supplier Med Vet International Supplier No. JOR322A1
JorVet Dip Quick Stain is excellent for use in blood cell differential counts or as a general cytology stain, and gives coloration similar to the standard Wright-Giemsa stain. The rapid 3-step procedure can be done right in the wide-mouth 180ml. container in 15 seconds. Fixative Only. 500mlNOTE: These components may interchange with other brands of 3-component differenential stains