Learn More
BURKLE INC Dip Sampler, PP transparent, volume 9.47 oz. (280 ml), 39.37 in. (100 cm)
Catalog No. 5000025685
Encompass_Preferred
High-quality dip sampler for efficient and reliable liquid sampling. Ideal for taking liquid samples in laboratories, industry, and water treatment. The transparent tube ensures an excellent view of the sample at all times. Equipped with a reflux valve at the upper end and a ball valve at the lower end for precise and controlled sampling.
Catalog No. 50-000-25685 Supplier BURKLE INC Supplier No. 53150100
High-quality dip sampler for efficient and reliable liquid sampling. Ideal for taking liquid samples in laboratories, industry, and water treatment. The transparent tube ensures an excellent view of the sample at all times. Equipped with a reflux valve at the upper end and a ball valve at the lower end for precise and controlled sampling.