Learn More
BURKLE INC Dip Sampler, PP, volume 16.91 oz. (500 ml), length 39.37-78.74 in. (100-200 cm)
Catalog No. 5000026032
Encompass_Preferred
High-quality dip sampler for efficient and reliable liquid sampling. Ideal for taking liquid samples in laboratories, industry, and water treatment. The transparent tube ensures an excellent view of the sample at all times. Equipped with a reflux valve at the upper end and a ball valve at the lower end for precise and controlled sampling.
Catalog No. 50-000-26032 Supplier BURKLE INC Supplier No. 53150200
High-quality dip sampler for efficient and reliable liquid sampling. Ideal for taking liquid samples in laboratories, industry, and water treatment. The transparent tube ensures an excellent view of the sample at all times. Equipped with a reflux valve at the upper end and a ball valve at the lower end for precise and controlled sampling.