Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Dip Stain Kits (Wright and Wright Giemsa)

Catalog No. ES2530
Change view
Click to view available options
Quantity:
4 x 3800 mL (1 gal.)
4 x 950 mL
Product Type:
Wright Dip Stain Kit
Wright Giemsa Dip Stain Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ES2530 4 x 3800 mL (1 gal.) Wright Giemsa Dip Stain Kit
ES2500 4 x 950 mL Wright Dip Stain Kit
ES2510 4 x 3800 mL (1 gal.) Wright Dip Stain Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES2530 Supplier ENG Scientific Supplier No. 2530
Add to Cart
Add to Cart

For Hemotology staining using Dip Stain Kit (Wright & Wright Giemsa). Kit includes Solution I - Fixative, Solution II - Wright or Wright Giemsa Dip Stain, Solution III - Matching Buffer, and Solution IV - Alcohol Wash.

  • Solutions made from certified dyes
  • Solutions should be replenished during staining period
  • During cold weather, allow solutions to return to room temperature and shake well before use
  • Dip Stain Kit includes: Wright or Wright Giemsa Dip Stain, methanol, buffers, stabilizers
  • Dip Stain Kit Replacement Solutions include: 1 gallon of solution
  • For in-vitro diagnostic use only

Specifications

Quantity 4 x 3800 mL (1 gal.)
Storage Requirements Room Temperature
For Use With (Application) For in-vitro diagnostic use only
Packaging Complete Kit (1 each Solutions I - IV)
Product Type Wright Giemsa Dip Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.