Learn More
Chemglass Life Sciences Dip Tube Assembly, 1/2" Sanitary, M18 x 1.5 Thread
SDP
Catalog No. NC2228477
Encompass
316L stainless steel diptube assembly with 1/2¿ sanitary fitting on top and fits into an 18mm head port plate.The length inside the bioreactor is 7.66¿.The overall length is 9.87¿.Electropolished Ra20 or better.
This item is not returnable. View return policy
Catalog No. NC2228477 Supplier Chemglass Life Sciences Supplier No. CLS138075
316L stainless steel diptube assembly with 1/2¿ sanitary fitting on top and fits into an 18mm head port plate.The length inside the bioreactor is 7.66¿.The overall length is 9.87¿.Electropolished Ra20 or better.