Learn More
Aviva Systems Biology Corp DIP2A Antibody - N-terminal region (ARP45852_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45852_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DIP2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DIP2A. Alternate names: C21orf106, DIP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59151