Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ DIPak™ Cartridge

Catalog No. p-3595242 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Cartridge
DI-PAK Purification Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
CPDI0NJM1 Cartridge
CPDI000L1 DI-PAK Purification Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CPDI0NJM1 Supplier MilliporeSigma™ Supplier No. CPDI0NJM1
Only null left
Add to Cart
Add to Cart

Produces deionized water directly from tap water

  • “Ion-exchange” and “depth filtration” technology
  • Produces 3–5 L of pure water per day
  • Feed water nature: Tap water
  • Can be easily snapped in and out in seconds without risk of leakage

TRUSTED_SUSTAINABILITY

Specifications

Feedwater Source Tap
Purification Method Ion Exchange, Depth Filtration
For Use With (Equipment) Milli-DI system
Type Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.